Learn More
Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate) (50 mg/mL)
Produced by bacterium Micromonospora rhodorangea and acts by binding ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells
Brand: Gibco™ 10131035
179.80 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
Gibco Geneticin Selective Antibiotic (G418 Sulfate) is produced by the bacterium Micromonospora rhodorangea and acts by binding the ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells.
Resistance to Geneticin Selection Antibiotic (G418 Sulfate) is conferred by the E. coli APH(3)–I and APH(3)–II resistance genes.
- Used as selective antibiotic in concentration range of 100 - 200μg/mL for bacteria, or 200 – 500μg/mL for most mammalian cells
- Supplied as 50mg/mL solution in water
Cell Culture, Cloning, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Selection, Sf9 & Sf21 Cell Culture, Transfection, Transformation
Order Info
Shipping Condition: Room Temperature
Compliance
Manufactured at a cGMP compliant facility, which is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Storage conditions: 2 to 8°C Shipping conditions: Ambient Shelf life: 24 months from date of manufacture |
|
50 mg/mL | |
Liquid | |
Sterile-filtered | |
Eukaryotic Selection/Stable Cell Line Generation | |
20 mL |
Eukaryotic Cells, Prokaryotic Cells | |
Mammalian Cell Culture, Insect Cell Culture | |
Antibiotic | |
Sterile-filtered | |
Geneticin™ | |
Room Temperature |
Safety and Handling
- Geneticin Selective Antibiotic (G418 Sulfate) (50 mg/mL)
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Gentamicin
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.