Learn More
Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate), Powder
Produced by bacterium Micromonospora rhodorangea and acts by binding the ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells
Brand: Gibco™ 11811023
87.68 GBP valid until 2024-12-31
Use promo code "21971" to get your promotional price.
Description
Resistance to Geneticin™ Selective Antibiotic (G418 Sulfate) is conferred by the E. coli APH (3) – I and APH (3) – II resistance genes.
- Used as selective antibiotic in concentration range of 100 - 200μg/mL for bacteria, or 200 – 500μg/mL for most mammalian cells
- Supplied as powder and should be made into a stock solution of 10 – 50mg/mL in water
Cell Culture, Cloning, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Selection, Sf9 & Sf21 Cell Culture, Transfection, Transformation
Order Info
Shipping Condition: Room Temperature
Compliance
Manufactured at a cGMP compliant facility, which is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Storage conditions: 15 to 30°C Shipping conditions: Ambient Shelf life: 24 months from date of manufacture |
|
10 to 50 μg/mL | |
Antibiotic | |
Eukaryotic Selection⁄Stable Cell Line Generation | |
1 g |
Eukaryotic Cells, Prokaryotic Cells | |
Powder | |
Sterile | |
Geneticin™ | |
Room Temperature |
Safety and Handling
- GENETICIN
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : D-Streptamine, O-2-amino-2,7-dideoxy-D-glycero-.alp ha.-D-gluco-heptopyranosyl-(1.fwdarw. 4)-O-[3-de
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.