missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate), Powder

Product Code. 11528676
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11528676

Brand: Gibco™ 11811031

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Produced by bacterium Micromonospora rhodorangea and acts by binding the ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells

Geneticin™ Selective Antibiotic (G418 Sulfate) is produced by the bacterium Micromonospora rhodorangea and acts by binding the ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells. Resistance to Geneticin™ Selective Antibiotic (G418 Sulfate) is conferred by the E. coli APH (3') – I and APH (3') – II resistance genes. Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate) is used as a selective antibiotic in the concentration range of 100 - 200 μg/mL for bacteria, or 200 – 500 μg/mL for most mammalian cells. This product is supplied as a powder and should be made into a stock solution of 10 – 50 mg/mL in water. We offer a variety of cell culture antibiotics for your convenience.

Product Use

For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

Dual site cGMP Manufacturing and Quality System

For supply chain continuity, we manufacture Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate) at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements, are certified to ISO 13485, and are registered with the FDA as medical device manufacturers.

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured at a cGMP compliant facility, which is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 15 to 30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Cell Type Eukaryotic Cells, Prokaryotic Cells
Concentration 10 to 50 μg/mL
Culture Type Mammalian Cell Culture, Insect Cell Culture
Form Powder
Product Type Antibiotic
For Use With (Application) Eukaryotic Selection/Stable Cell Line Generation
Product Line Geneticin
Quantity 5 g
Shipping Condition Room Temperature
compliance-icons
Product Identifier
  • Geneticin Selective Antibiotic (G418 Sulfate), Powder
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : O-2-amino-2,7-dideoxy-D-glycero-.alpha.-D-gluco-heptopyranosyl-(1.fwdarw.4)-O-[3-deoxy-4-C-methyl-3-

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.