missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Blasticidin S HCl (10 mg/mL)

Product Code. 12172530
Click to view available options
Quantity:
20 mL
10 x 1 mL
Unit Size:
10mL
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 12172530

Brand: Gibco™ A1113903

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Selective antibiotic in concentration range of 50G–100μg/mL

Blasticidin S is a nucleoside antibiotic isolated from Streptomyces griseochromogenes. It is a potent inhibitor of protein synthesis in both prokaryotic and eukaryotic cells. Blasticidin S acts quickly by inhibiting peptide bond formation by the ribosome.

  • Used for prevention of contamination of cell cultures
  • Recommended working concentration ranges 1–20μg/mL depending on cell line
  • Provided as solution at 10mg/mL in 20 mM HEPES (pH 7.2–7.5)
  • Supplied in 10 vials each containing 1mL.
  • Resistance is conferred by expression of either one of two Blasticidin S deaminase genes: BSD or bsr

Cell Culture, Cloning, Drosophila S2 Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Selection, Sf9 and Sf21 Cell Culture, Transfection, Transformation

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: -5°C to -20°C (protect from light)
Shipping conditions: Dry ice
Shelf life: 9 months from date of manufacture
Cell Type Eukaryotic Cells, Prokaryotic Cells
Concentration 10 mg/mL
Culture Type Mammalian Cell Culture, Insect Cell Culture
Form Liquid
Product Type Antibiotic
Sterility Sterile-filtered
With Additives HEPES
Product Line Gibco
Quantity 10 x 1 mL
compliance-icons
Product Identifier
  • Blasticidin S HCL
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contains : Blasticidin S HCL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.