missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Puromycin Dihydrochloride

Aminonucleoside antibiotic produced by Streptomyces alboniger

Brand:  Gibco™ A1113802

427.65 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10296974

  • £514.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Puromycin works by inhibiting peptidyl transfer on both prokaryotic and eukaryotic ribosomes; resistance is conferred by the expression of the pac gene.

  • Used in cell biology as selection antibiotic in mammalian cell culture systems
  • Recommended working concentration ranges from 0.2–5.0μg/mL
  • Can be toxic to eukaryotic cells at concentrations as low as 1μg/mL
  • Supplied at 10mg/mL in 20 mM HEPES buffer (pH 7.2–7.5)

Cell Culture, Cloning, Drosophila S2 Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Selection, Sf9 & Sf21 Cell Culture, Transfection, Transformation

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: -5 to -20°C
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture
10 mg/mL
Liquid
Sterile-filtered
Eukaryotic Selection⁄Stable Cell Line Generation
1 bottLe
Eukaryotic Cells, Prokaryotic Cells
Mammalian Cell Culture, Insect Cell Culture
Antibiotic
HEPES
Gibco™
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.