missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Puromycin Dihydrochloride

Product Code. 12122530
Click to view available options
Quantity:
20 mL
10 x 1 mL
Unit Size:
10mL
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 12122530

Brand: Gibco™ A1113803

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Aminonucleoside antibiotic produced by Streptomyces alboniger

Puromycin works by inhibiting peptidyl transfer on both prokaryotic and eukaryotic ribosomes; resistance is conferred by the expression of the pac gene.

  • Used in cell biology as selection antibiotic in mammalian cell culture systems
  • Recommended working concentration ranges from 0.2–5.0μg/mL
  • Can be toxic to eukaryotic cells at concentrations as low as 1μg/mL
  • Supplied at 10mg/mL in 20 mM HEPES buffer (pH 7.2–7.5)

Cell Culture, Cloning, Drosophila S2 Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Selection, Sf9 & Sf21 Cell Culture, Transfection, Transformation

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: -5 to -20°C
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture
Concentration 10 mg/mL
Culture Type Mammalian Cell Culture, Insect Cell Culture
Form Liquid
Product Type Antibiotic
Sterility Sterile-filtered
With Additives HEPES
For Use With (Application) Eukaryotic Selection⁄Stable Cell Line Generation
Product Line Gibco
Quantity 10 x 1 mL
Shipping Condition Dry Ice
Show More Show Less
compliance-icons
Product Identifier
  • Puromycin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : puromycin

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.