missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ GlutaMAX™ Supplement
GREENER_CHOICE

Product Code. 13462629
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 13462629

Brand: Gibco™ 35050061

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GlutaMAX Supplement is an L-alanyl-L-glutamine dipeptide alternative to L-glutamine. It exhibits increased stability over L-glutamine leading to improved cellular health.

Gibco GlutaMAX Supplement is an L-alanyl-L-glutamine dipeptide alternative to L-glutamine that exhibits increased stability, leading to improved cell health. L-Glutamine is an amino acid required in cell culture for the formation of purine and pyrimidine nucleotides, amino sugars, glutathione, L-glutamate, and other amino acids, as well as in protein synthesis and glucose production. GlutaMAX Supplement is suitable for adherent and suspension culture of mammalian cells, with no adaptation required. It is formulated as a 200 mM L-alanyl-L-glutamine dipeptide in 0.85% NaCl.

Compared to L-glutamine, GlutaMAX Supplement:

  • Minimizes toxic ammonia build-up
  • Improves cell viability and growth
  • Remains stable across a wide range of temperatures

Minimizes toxic ammonia build-up

Unlike L-glutamine, GlutaMAX Supplement does not spontaneously break down to form ammonia. Instead, cells cleave the dipeptide bond to release L-glutamine as needed. This system prevents waste build-up and maintains a fresh supply of L-glutamine during long-term culture.

Improves cell viability and growth

Cultures with lower levels of ammonia show improved cell viability. In addition, culture conditions are more controlled due to the consistent concentration of GlutaMAX Supplement that is not possible with L-glutamine.

Remains stable across a wide range of temperatures

GlutaMAX Supplement is room-temperature stable and ready to use. It retains full functionality for 24 months at room temperature, 2 to 8°C, or -5 to -30°C for convenient storage and handling. GlutaMAX Supplement is even stable at 37°C for a week or more.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 15°C to 30°C
Alternate storage conditions: -5°C to -20°C and 2°C to 8°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture
Cell Type Mammalian Cells
Classification Animal Origin-free
Concentration 100X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type GlutaMax
Sterility Sterile-filtered
For Use With (Application) Clinical Research, Translational Research
Green Features Sustainable packaging
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Osmolality 430 to 490 mOsm/kg
pH 5 to 6
Product Line Gibco, GlutaMAX
Quantity 100 mL
Shipping Condition Room Temperature
Source Animal Origin-free
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.