Learn More
Gibco™ Grace's Insect Medium (2X), supplemented
For use in the agarose overlay when performing the plaque assay
Brand: Gibco™ 11667037
Code : 50
Additional Details : Weight : 0.14000kg
Description
Grace's Supplemented Insect Medium was originally designed for the growth of cells from the Australian Emperor Gum Moth, Antherea eucalypti. Used for the growth of Spodeptera frugiperda cells, Sf9 and Sf21. Sf9 and Sf21 cells are routinely used to express recombinant protein using the Baculovirus Expression Vector System (BEVS). Modified as follows: With Lactalbumin hydrolysate, Yeastolate, L-glutamine and Sodium bicarbonate. The complete formulation is available. Grace's Supplemented Insect Medium, 2x contains lactalbumin hydrolysate and yeastolate not commonly found in the unsupplemented version commonly diluted with sterile water, Fetal bovine Serum and agarose before use in the agarose overlay, as described in the product manual.
Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture
Order Info
Shipping Condition: Room temperature
Compliance
Gibco Grace's Supplemented Insect Medium, 2x is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Storage conditions: 2-8°C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Insect Cell | |
Grace's Insect Medium | |
Sustainable packaging | |
Room Temperature |
Sf21, Sf9 | |
Liquid | |
Glutamine, Lactalbumin Hydrolysate, Yeastolate | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.