missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Grace's Insect Medium, supplemented
Widely used for the growth of Spodeptera frugiperda cells, Sf9 and Sf21
£76.25
Specifications
Content And Storage | Storage conditions: 2°C to 8°C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
---|---|
Cell Line | Sf21, Sf9 |
Cell Type | Insect Cell |
Form | Liquid |
Product Type | Grace's Insect Medium |
Description
Originally designed for the growth of cells from the Australian Emperor Gum Moth, Antherea eucalypti. Sf9 and Sf21 cells are routinely used to express recombinant protein using the Baculovirus Expression Vector System (BEVS).
- Contains lactalbumin hydrolysate, yeastolate, L-glutamine, and sodium bicarbonate
- Dual site cGMP manufacturing and quality system
- Commonly supplemented with 10% Fetal Bovine Serum (FBS) before use
Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture
Order Info
Shipping Condition: Room temperature
Specifications
Storage conditions: 2°C to 8°C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Insect Cell | |
Grace's Insect Medium | |
No Insect Hemolymph | |
Calcium, Magnesium | |
Room Temperature |
Sf21, Sf9 | |
Liquid | |
Glutamine, Yeastolate, Lactalbumin Hydrolysate | |
Sustainable packaging | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.