Learn More
Gibco™ Gurr Buffer Tablets
A convenient way to prepare a pH 6.8 phosphate buffer for dilution of Giemsa stain
Brand: Gibco™ 10582013
76.71 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 0.05000kg
Description
- Used in G-banding of chromosomes for cytogenetic analysis
- Add one buffer tablet to 100 mL of distilled water and stir until dissolved
- Package size: 50 tablets
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Room Temperature
Specifications
Storage conditions: 15–30°C Shipping conditions: Ambient Shelf life: 24 months from date of manufacture |
|
Powder | |
Clinical Research, Translational Research | |
50 TabLets |
Mammalian Cells | |
Gurr Buffer | |
6.8 | |
Room Temperature |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.