missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Ham's F-12 Nutrient Mix
GREENER_CHOICE

Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free, single-cell plating of Chinese Hamster Ovary (CHO) cells.

Brand:  Gibco™ 11765054

26.64 GBP valid until 2025-12-31
Use promo code "24273" to get your promotional price.


 View more versions of this product

Product Code. 15172529

  • £32.38 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Requires : Supplementation
This item is not returnable. View return policy

Description

Description

Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free, single-cell plating of Chinese Hamster Ovary (CHO) cells. F-12 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including chondrocytes and rat prostate epithelial cells. We offer a variety of F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This Ham's F-12 is modified as follows:

With: L-glutamine, Phenol Red

The complete formulation is available.

Using F-12

Compared to other basal media, F-12 contains a wider variety of components, including zinc, putrescine, hypoxanthine, and thymidine. F-12 contains no proteins or growth factors. Therefore, F-12 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-12 uses a sodium bicarbonate buffer system (1.176 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2°C to 8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Primary Rat Astrocytes
1 X
Ham's F-12 Nutrient Mixture
Sterile-filtered
No HEPES
Gibco™
Room Temperature
CHO, COS-7, and rat prostate epithelial cells
Animal Origin-free
Liquid
Sterile-filtered
Glutamine, Phenol Red, Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2472346 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11765054
Lot Number2537750 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11765054
Lot Number2522685 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11765054
Lot Number2618879 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11765047,11765054
Lot Number2537116 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11765047,11765054
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Ham's F-12 Nutrient Mix > Quantity: 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.