Learn More
Gibco™ Ham's F-12 Nutrient Mix
Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free, single-cell plating of Chinese Hamster Ovary (CHO) cells.
£23.48 - £30.89
Specifications
Cell Line | CHO, COS-7, and rat prostate epithelial cells |
---|---|
Cell Type | Primary Rat Astrocytes |
Classification | Animal Origin-free |
Concentration | 1 X |
Form | Liquid |
Requires: Supplementation
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15172529
|
Gibco™
11765054 |
500 mL |
£30.89
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15440564
|
Gibco™
11765047 |
1000 mL |
£23.48
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free, single-cell plating of Chinese Hamster Ovary (CHO) cells. F-12 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including chondrocytes and rat prostate epithelial cells. We offer a variety of F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This Ham's F-12 is modified as follows:
With: L-glutamine, Phenol Red
The complete formulation is available.
Using F-12
Compared to other basal media, F-12 contains a wider variety of components, including zinc, putrescine, hypoxanthine, and thymidine. F-12 contains no proteins or growth factors. Therefore, F-12 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-12 uses a sodium bicarbonate buffer system (1.176 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Specifications
CHO, COS-7, and rat prostate epithelial cells | |
Animal Origin-free | |
Liquid | |
Sterile-filtered | |
No HEPES | |
Gibco™ |
Primary Rat Astrocytes | |
1 X | |
Ham's F-12 Nutrient Mixture | |
Glutamine, Phenol Red, Sodium Pyruvate | |
cGMP-compliant under the ISO 13485 standard |
Safety and Handling
missing translation for 'shelfLife' : 12 Months From Date of Manufacture
Your input is important to us. Please complete this form to provide feedback related to the content on this product.