missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Ham's F-12K (Kaighn's) Medium
GREENER_CHOICE

Product Code. 11580556
Click to view available options
Quantity:
500 mL
10 x 500 mL
Unit Size:
500mL
Pack of 10
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11580556

Brand: Gibco™ 21127022

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Ham's F-12K (Kaighn's) Medium is a modification of Ham's F-12 Nutrient Mixture. Ham's F-12K (Kaighn's) Medium was developed for primary human hepatocytes, as well as for some rat and chicken liver cells in a reduced serum environment.

Ham's F-12K (Kaighn's) Medium is a modification of Ham's F-12 Nutrient Mixture. Ham's F-12K (Kaighn's) Medium was developed for primary human hepatocytes, as well as for some rat and chicken liver cells in a reduced serum environment.

This F-12K (Kaighn's) Medium is manufactured as follows:

With: L-glutamine, Phenol Red

Without: HEPES

The complete formulation is available.

Using Ham's F-12K

Ham's F-12K (Kaighn's) Medium contains many components not found in traditional basal media, such as putrescine, thymidine, hypoxanthine, zinc, and higher levels of all amino acids and sodium pyruvate. These additions allow the medium to be supplemented with very low levels of serum or defined components, for some cell types. Ham's F-12K (Kaighn's) Medium contains no proteins or growth factors, and is therefore often supplemented with growth factors and Fetal Bovine Serum (FBS). The FBS concentration must be optimized for each cell line. Ham's F-12K (Kaighn's) Medium uses a sodium bicarbonate buffer system (2.5 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Human Hepatocytes
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type Ham's F-12K (Kaighn's) Medium
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Low Glucose, Glutamine, Phenol Red, Sodium Pyruvate
Without Additives No HEPES
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Silver chloride
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P390-Absorb spillage to prevent material damage.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Silver chloride

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.