Learn More
Gibco™ HBSS (10X), calcium, magnesium, no phenol red
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
Brand: Gibco™ 14065056
40.05 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media, or for reagent preparation. Concentrated forms of Gibco™ HBSS require sodium bicarbonate addition, pH adjustment, and filtration at the time of preparation (see protocol for details).
We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.
This HBSS is modified as follows:
With: Calcium, Magnesium, Glucose
Without: Phenol Red, Sodium Bicarbonate
The complete formulation is available.
Order Info
Shipping Condition: Room Temperature
Specifications
No Sodium Pyruvate | |
Storage conditions: 15°C to 30°C Shipping conditions: Room temperature Shelf life: 24 months from date of manufacture |
|
Gibco™ | |
500 mL | |
Variety of Cell Culture Applications; Such as Washing Cells Before Dissociation; Transporting Cells or Tissue Samples; Diluting Cells for Counting And Preparing Reagents | |
4.4 - 4.7 | |
Room Temperature | |
Liquid |
Animal Origin-Free | |
10 X | |
Hanks' Balanced Salt Solution (HBSS) | |
Hank's Balanced Salt | |
2700 - 3070 mOsm/kg | |
No Phenol Red | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.