Learn More
Gibco™ HBSS, calcium, magnesium
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.
Brand: Gibco™ 24020117
15.58 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
This HBSS is modified as follows:
With: Calcium, Magnesium, Glucose, Phenol Red
The complete formulation is available.
Order Info
Shipping Condition: Room Temperature
Specifications
No Sodium Pyruvate | |
Storage conditions: 15-30° C Shipping conditions: Ambient Shelf life: 36 months from date of manufacture |
|
Gibco™ | |
500 mL | |
Variety of Cell Culture Applications; Such as Washing Cells Before Dissociation; Transporting Cells or Tissue Samples; Diluting Cells for Counting; and Preparing Reagents | |
6.7 - 7.8 | |
Room Temperature | |
Liquid |
Animal Origin-Free | |
1 X | |
Hanks' Balanced Salt Solution (HBSS) | |
Hank's Balanced Salt | |
270 - 305 mOsm/kg | |
Phenol Red | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.