Learn More
Gibco™ HBSS, calcium, magnesium, no phenol red
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
£23.41
Spécification
Additive | No Sodium Pyruvate |
---|---|
Classification | Animal Origin-Free |
Dilution | 1 X |
Product Line | Gibco™ |
Product Type | Hanks' Balanced Salt Solution (HBSS) |
Description
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media, or for reagent preparation.
We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.
This HBSS is modified as follows:
With: Calcium, Magnesium, Glucose
Without: Phenol Red, Sodium Pyruvate
The complete formulation is available.
Informations sur la commande
Shipping Condition: Room Temperature
Spécification
No Sodium Pyruvate | |
1 X | |
Hanks' Balanced Salt Solution (HBSS) | |
Mammalian Cell Culture | |
270 - 305 mOsm/kg | |
No Phenol Red | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
Animal Origin-Free | |
Gibco™ | |
Hank's Balanced Salt | |
Sustainable packaging | |
6.7 - 7.8 | |
Room Temperature | |
Liquid |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.