missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Horse Serum, heat inactivated, New Zealand origin

Product Code. 10368902
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10368902

Brand: Gibco™ 26050088

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

EIA tested (negative) serum from a donor herd

  • Gibco sera offers excellent value for basic cell culture, specialty research, and specific assays
  • It has earned the trust of researchers with consistent quality and award-winning support
  • Helps meet research needs and budget requirements
  • EIA tested (negative) serum from a donor herd
  • Shipped frozen, store at <10°C
  • Sera Category: Specialty
  • Origin: New Zealand

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: ≤-10°C, Shipping conditions: Frozen
Species Horse/Equine, Horse
Country of Origin New Zealand
Form Liquid
Product Type Horse Serum
Quantity 500 mL
Serum Treatment Heat-inactivated
Product Line Gibco
Shipping Condition Frozen

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.