Learn More
Gibco™ Horse Serum, New Zealand origin
Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support.
Brand: Gibco™ 16050122
62.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Code : 50
Additional Details : Weight : 0.61000kg
Description
Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera Category: Specialty
- Origin: New Zealand
- EIA tested (negative) serum from a donor herd
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Horse/Equine, Horse | |
Liquid | |
500 mL | |
Sterile | |
Frozen |
Adult | |
New Zealand | |
Horse Serum | |
Standard (Sterile-filtered) | |
Sustainable packaging |
For In Vitro Diagnostic Use.