Learn More
Gibco™ Hygromycin B (50 mg/mL)
Water-soluble antibiotic purified from bacterium Streptomyces hydroscopicus
Brand: Gibco™ 10687010
174.64 GBP valid until 2025-12-31
Use promo code "24361" to get your promotional price.
Description
Hygromycin B is a water-soluble antibiotic purified from the bacterium Streptomyces hydroscopicus. Gibco™ Hygromycin B is used as a bacterial selection antibiotic in the concentration range of 200–500 μg/mL. This product is supplied as a 50 mg/mL solution.
Resistance to Hygromycin B is conferred by the E. coli hygromycin resistance gene (hyg or hph). Hygromycin B has a different mode of action than Geneticin™, Blasticidin S, or Zeocin™ selection reagents, making it an excellent choice for dual-selection experiments.
Order Info
Shipping Condition: Room Temperature

Specifications
Storage conditions: 2 to 8°C Shipping conditions: Ambient |
|
Mammalian Cell Culture, Insect Cell Culture | |
Antibiotic | |
Eukaryotic Selection⁄Stable Cell Line Generation | |
Room Temperature |
50 mg/mL | |
Liquid | |
Sterile | |
20 mL |
Safety and Handling
- HYGROMYCIN B
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
Hazard Statement
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
Supplemental information
- MIXTURE LIST-Contains : Hygromycin B
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
H044-149US | Certificate of Analysis | 31/01/2025 | 10687010 |
H044-155US | Certificate of Analysis | 31/01/2025 | 10687010 |
H044-154US | Certificate of Analysis | 31/01/2025 | 10687010 |
H044-152US | Certificate of Analysis | 31/01/2025 | 10687010 |
H044-153US | Certificate of Analysis | 31/01/2025 | 10687010 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.