Learn More
Gibco™ Human IL-10 Recombinant Protein
Description
Carrier-Free Reconstitution: Centrifuge the vial briefly, before opening to bring the contents to the bottom. Reconstitute the lyophilized protein in sterile water or other appropriate buffered solutions to 0.1-1.0 mg/mL. Apportion the reconstituted protein into working aliquots and store at ≤-20°C. Make any further dilutions of the reconstituted protein in solution with carrier protein, such as PBS with 1% BSA. Storage: Store the lyophilized protein at 2-8°C or -20°C long term, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤-20°C. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Accession Number | P22301 |
| For Use With (Application) | Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 3586 |
| Molecular Weight (g/mol) | 18.6 kDa |
| Name | Human IL-10 |
| Purification Method | Purified |
| Quantity | 5 μg |
| Storage Requirements | 4°C |
| Regulatory Status | RUO |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.