missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IMDM, GlutaMAX™ Supplement
GREENER_CHOICE

IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells.

£33.95

Specifications

Content And Storage Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Cell Line Jurkat, COS-7, and macrophage cells
Classification Animal Origin-free
Concentration 1 X
Form Liquid
View More Specs

Requires: Supplementation


Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12539089
GREENER_CHOICE
View Documents Promotion Details
Gibco™
31980030
500 mL
£33.95
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This IMDM is modified as follows:
With: GlutaMAX™, Phenol Red
Without: α-thioglycerol, 2-mercaptoethanol

The complete formulation is available.

Using IMDM
IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate. IMDM with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. IMDM contains no proteins, lipids, or growth factors. Therefore, IMDM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). IMDM uses a sodium bicarbonate buffer system (3.024 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Animal Origin-free
Liquid
Sterile-filtered
No α-Thioglycerol, No 2-Mercaptoethanol
cGMP-compliant under the ISO 13485 standard
Room Temperature
Jurkat, COS-7, and macrophage cells
1 X
IMDM (Iscove's Modified Dulbecco's Medium)
High Glucose, GlutaMAX, HEPES, Phenol Red, Sodium Pyruvate
Sustainable packaging
Gibco™, GlutaMAX™
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 Months From Date of Manufacture

SDS
Documents

Documents

Product Certifications

In vitro diagnostic