missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IMDM
GREENER_CHOICE

Product Code. 10135083
Click to view available options
Quantity:
500 mL
1000 mL
10 x 500 mL
Unit Size:
Each
Pack of 10
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10135083

Brand: Gibco™ 12440046

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Requires

Supplementation

IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells

IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This IMDM is modified as follows:

With: L-glutamine, Phenol Red

Without: α-thioglycerol, 2-mercaptoethanol

The complete formulation is available.

Using IMDM

IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate. IMDM contains no proteins, lipids, or growth factors. Therefore, IMDM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). IMDM uses a sodium bicarbonate buffer system (3.024 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Jurkat, COS-7, and macrophage cells
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type IMDM (Iscove's Modified Dulbecco's Medium)
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives High Glucose, Glutamine, HEPES, Phenol Red, Sodium Pyruvate
Without Additives No α-Thioglycerol, No 2-Mercaptoethanol
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 1000 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.