missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Insulin-Transferrin-Selenium-Ethanolamine (ITS -X) (100X)

A basal medium supplement to reduce the amount of fetal bovine serum (FBS) needed to culture cells

Brand:  Gibco™ 51500056

48.00 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 10524233

  • £60.00 / 10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

  • ITS-X solution is routinely used, in combination with 2 to 4% FBS, to support cell types that require ethanolamine, such as bronchial epithelial cell
  • Insulin promotes glucose and amino acid uptake, lipogenesis, intracellular transport, and the synthesis of proteins and nucleic acids
  • Transferrin is an iron carrier and may also help to reduce toxic levels of oxygen radicals and peroxide
  • Selenium, as sodium selenite is a cofactor for glutathione peroxidase and other proteins that is used as an antioxidant in media
  • Ethanolamine is a precursor of phosphoglycerides which are essential to the structure of the plasma membrane and cellular organelles
  • Manufactured at a cGMP compliant facility, located in Grand Island, New York, registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2°C to 8°C
Shipping conditions: Room temperature
Shelf life: 18 months from date of manufacture
100 X
Transferrin
Insulin
Room Temperature
Mammalian Cells
Liquid
Sterile-filtered
10 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.