missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Insulin-Transferrin-Selenium (ITS -G) (100X)
A basal medium supplement to reduce the amount of fetal bovine serum (FBS) needed to culture cells
Brand: Gibco™ 41400045
Code : 50
Additional Details : Weight : 0.02000kg
Description
- Originally used with RPMI 1640 and Minimum Essential Medium (MEM), ITS-G solution is now routinely used with other basal media to support a wide variety of cell types in the presence of 2 to 4% FBS
- Insulin promotes glucose and amino acid uptake, lipogenesis, intracellular transport, and the synthesis of proteins and nucleic acids
- Transferrin is an iron carrier and it may also help to reduce toxic levels of oxygen radicals and peroxide
- Selenium, as sodium selenite, is a cofactor for glutathione peroxidase and other proteins, and is used as an antioxidant in media
Order Info
Shipping Condition: Room Temperature
Specifications
Storage conditions: 2°C to 8°C Shipping conditions: Room temperature Shelf life: 18 months from date of manufacture |
|
100 X | |
Transferrin | |
No Phenol Red | |
Room Temperature |
Mammalian Cells | |
Liquid | |
Sterile-filtered | |
10 mL |
For Research Use Only. Not for use in diagnostic procedures.