missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Kanamycin Sulfate
GREENER_CHOICE

Product Code. p-4918406
Click to view available options
Quantity:
5 g
25 g
100 mL
Form:
Liquid
Powder
Unit Size:
100mL
25g
5g
This item is not returnable. View return policy

Product Code. 15470594

Brand: Gibco™ 15160054

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Water-soluble antibiotic originally purified from bacterium Streptomyces kanamyceticus

  • A water-soluble antibiotic originally purified from the bacterium Streptomyces kanamyceticus
  • Kanamycin acts by binding to the 30S subunit of the bacterial ribosome and inhibiting protein synthesis in susceptible bacteria
  • Gibco™ Kanamycin is effective against a wide variety of gram-negative and some gram-positive bacteria
  • It is used for the prevention of bacterial contamination of cell cultures
  • Provided as a 10 mg/mL stock solution, the recommended working concentration for cell culture is 100 μg/mL
  • Manufactured at a cGMP compliant facility that is registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards
  • Store at -5°C to -20°C

For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured at a cGMP compliant facility, which is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: -5 to -20°C
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture
Concentration 100 X, 10 mg/mL
Form Liquid
Product Type Antibiotic
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
For Use With (Application) Bacterial Selection
Quantity 100 mL
Shipping Condition Dry Ice
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity Category 1
Hazard Statement
  • H360-May damage fertility or the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Kanamycin sulphate

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.