Learn More
Gibco™ L-Glutamine (200 mM)

Ready-to-use stock solution that acts as a cell culture supplement
Brand: Gibco™ 25030081
9.95 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
L-Glutamine is an amino acid that is required for cell culture. L-Glutamine participates in the formation of purine and pyrimidine nucleotides, amino sugars, glutathione, L-glutamate, and other amino acids, as well as in protein synthesis and glucose production. Unlike most other amino acids, L-glutamine is not stable in solution. The rate at which degradation proceeds is a function of time, temperature, and pH.
- Optimal concentration is dependent upon the cell type and medium used to culture the cells, but generally falls in the range of 2 to 6 mM
- Manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK, registered with the FDA as a medical device manufacturer and certified to the ISO 13485 standard
Bioproduction, Cell Culture, Cloning, Mammalian Cell Culture, Transformation, Viral Vaccine Production
Order Info
Shipping Condition: Dry Ice

Specifications
Glutamine | |
100 mL | |
Liquid | |
Dry Ice |
5 to 6 | |
100X | |
Gibco™ | |
Sustainable packaging |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.