missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Lab Armor™ Beads
GREENER_CHOICE

Product Code. p-4921385
Click to view available options
Quantity:
Volume: 4 L
Volume: 8 L
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10120988

Brand: Gibco™ A1254301

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Small, non-uniform metal beads comprised of dry, metallic thermal alloy for use in water baths designed to replace water in a water bath or ice in an ice bucket.

  • Maintains uniform temperature better than water by transferring heat more slowly
  • Can remain "always-on" and ready to use
  • Bottles and tubes stay in place without need for racks, weights and floats
  • Removes need for water bath maintenance such as emptying, cleaning and refilling bath
  • Eliminates risk of contamination from standing water in water bath
  • No condensation seen around caps and incubating bottles removed from dry bath
  • Environmentally responsible alternative to water
  • More energy efficient than water
  • Draws less energy when run at 37 and 65°C
  • Made from recyclable material that can be cleaned with ethanol, eliminating need for water bath germicides that can harm the environment
  • Store at 15 to 30°C (shipped under ambient conditions)
TRUSTED_SUSTAINABILITY

Specifications

Bead Type Reflective, Inert, Smooth
Content And Storage Storage conditions: 15 – 30°C
Shipping conditions: Ambient
Capacity (Metric) 4 L
Temperature Range -80°C to +180°C
Type Bead
Green Features Energy efficient
Product Line Gibco
Quantity Volume: 4 L
Shipping Condition Room Temperature
Diameter (Metric) 5 mm poly-disperse
compliance-icons
Product Identifier
  • 4-Nitrobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : 4-Nitrobenzoyl chloride

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.