Learn More
Gibco™ Sheep Serum, New Zealand origin
Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
Brand: Gibco™ 16070096
Code : 50
Additional Details : Weight : 0.50000kg
Description
Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
- Sera Category: Specialty
- Origin: New Zealand
Order Info
Shipping condition: Dry ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
New Zealand | |
Sheep Serum | |
Standard (Sterile-filtered) | |
Sustainable packaging | |
Frozen |
Sheep/Ovine | |
Liquid | |
500 mL | |
Sterile-filtered | |
Gibco™ |
For In Vitro Diagnostic Use.