Learn More
Gibco™ Leibovitz's L-15 Medium, powder
Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue.
£50.75
Specifications
Content And Storage | Storage conditions: 2-8° C Shipping conditions: Ambient Shelf life: 36 months from date of manufacture |
---|---|
Cell Line | HEP-2 monkey kidney cells |
Cell Type | Explants of embryonic and adult human tissue |
Classification | Animal Origin |
Form | Powder |
Requires: Supplementation
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11504486
|
Gibco™
41300070 |
10 L |
£50.75
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue. L-15 is buffered by phosphates and free base amino acids instead of sodium bicarbonate. This medium is designed for supporting cell growth in environments without CO2 equilibration.
This L-15 is modified as follows:
With: Galactose, Phenol Red, L-glutamine, Sodium Pyruvate
Without: Glucose, HEPES, Sodium Bicarbonate
The complete formulation is available.
L-15 contains no proteins, lipids, or growth factors. Therefore, F-10 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). Powder forms of Gibco™ L-15 cell culture medium require pH adjustment and filtration at the time of preparation (see protocol for details).
Order Info
Shipping Condition: Room Temperature
Specifications
Storage conditions: 2-8° C Shipping conditions: Ambient Shelf life: 36 months from date of manufacture |
|
Explants of embryonic and adult human tissue | |
Powder | |
Sterile-filtered | |
No Glucose, No HEPES, No Sodium Bicarbonate | |
Gibco™ |
HEP-2 monkey kidney cells | |
Animal Origin | |
Leibovitz's L-15 Medium | |
Glutamine, Phenol Red, Sodium Pyruvate, Galactose | |
cGMP-compliant under the ISO 13485 standard | |
Room Temperature |
Safety and Handling
missing translation for 'shelfLife' : 36 Months From Date of Manufacture
Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. For in vitro diagnostic use.