Learn More
Gibco™ LV-MAX™ Lentiviral Production System
Brand:  Gibco™ A35684
Product Code. 15829081
  • £2195.00 / 1 set
Explore more special offers
Description
Description
The Gibco LV-MAX Lentiviral Production System is the first optimized system that provides a scalable and high-yield lentiviral vector production platform. It is based on a suspension, high-density culture of HEK 293-derived Viral Production Cells adapted to a special chemically defined, serum-free and protein-free LV-MAX Production Medium. Optimal viral titer yield is mediated by our advanced lipid nanoparticle transfection reagent in combination with a novel lentiviral-specific enhancer and supplement. All components work synergistically to generate superior and functional lentiviral particles under serum-free conditions compared to conventional polyethylenimine (PEI).
The LV-MAX Lentiviral Production System provides:
  • High titer—greater than 1 x 108 TU/mL (unconcentrated LVVs-GFP)
  • Scalable suspension system—from 96 deep-well plate to 2-L bioreactor
  • Serum-free—eliminating animal-origin components and associated risks
  • Easy, robust culture and transfection protocols—fits into current transient production workflows
The components included in the LV-MAX Lentiviral Production System are: two vials of frozen Viral Production Cells, one liter of LV-MAX Production Medium, and one LV-MAX Transfection Kit sufficient to produce 0.3 L of lentivirus. All components of the LV-MAX Lentiviral Production System are also available for purchase separately. Also required, but not included in this kit, is Gibco Opti-MEM I Reduced Serum Medium (Cat. No. 31985062).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
• 1 L LV-MAX Production Medium. Store at 2–8°C, protect from light.
• 0.3 L LV-MAX Transfection Kit. Store at 2–8°C, protect from light.
• 2 vials of Viral Production Cells. Store in liquid nitrogen until ready to use. Do not store the cells at -80°C.
Viral Production Cell-Line
Transient
Lentiviral Production System
1 Kit
Lentiviral, Transfection
Suspension Cell Culture
Liquid
LV-MAX™
Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Gibco™ LV-MAX™ Lentiviral Production System > 1 kit