Learn More
Gibco™ LV-MAX™ Production Medium
Description
Gibco LV-MAX Production Medium is a chemically defined, serum-free, protein-free medium, specially developed for high-density growth and transfection of Gibco Viral Production Cells (suspension-adapted HEK 293-derived cells) promoting high-titer lentiviral production.
- Supports growth of suspension Viral Production Cells to densities of over 1.0 x 107 cells/mL
- Enables sustained, high-level production of high-density transiently transfected cultures, achieving titer of up to 1 x 108 TU/mL (unconcentrated LVVs-GFP)
- Supports growth and transfection of Viral Production Cells in culture formats from 96-deep well blocks to 2-L flasks and bioreactors
LV-MAC Production Medium is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. The medium is not recommended for adherent cell cultures.
Specifications
Specifications
| Cell Line | Viral Production Cells |
| Cell Type | HEK 293 |
| Classification | Animal Origin-free, Chemically-defined, Protein-free, Serum-free |
| Form | Liquid |
| Product Type | Lentiviral Production Medium |
| Sterility | Sterile-filtered |
| With Additives | GlutaMAX |
| Manufacturing Quality | ISO 13485, MDSAP, FDA-registered |
| Product Line | LV-MAX |
| Quantity | 6 x 1 L |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.