missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ McCoy's 5A (Modified) Medium
GREENER_CHOICE

McCoy's 5A (modified) Medium is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues.

Brand:  Gibco™ 16600082

30.64 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.


 View more versions of this product

Product Code. 15410604

  • £35.31 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Requires : Supplementation
This item is not returnable. View return policy

Description

Description

McCoy's 5A (modified) Medium is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues. This medium will support the growth of primary mammalian cells derived from normal bone marrow, skin, spleen, kidney, lung, rat embryos, and other tissues.

This McCoy's 5A is modified as follows:

With: High Glucose, L-glutamine, Bacto-peptone, Phenol Red

Without: Sodium Pyruvate, HEPES

The complete formulation is available.

Using McCoy's 5A (modified) Medium

Dr. Thomas McCoy originally formulated McCoy's 5A medium as a modification of Basal Medium 5A. Unlike other media, McCoy's 5A contains the reducing agent glutathione, bacto-peptone, and a high level of glucose. This product also includes Dr. Hsu's addition of Hanks' salts to enable use outside a CO2 incubator. McCoy's 5A (modified) Medium requires serum supplementation, commonly with 10% Fetal Bovine Serum (FBS). McCoy's 5A (modified) Medium uses a sodium bicarbonate buffer system (2.2 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Biopsy Tissues
1 X
McCoy's 5A Modified Medium
Sterile-filtered
No HEPES, No Sodium Pyruvate
Gibco™
Room Temperature
Rat fibroblasts
Animal Origin
Liquid
Sterile-filtered
High Glucose, Glutamine, Phenol Red, Sodium Bicarbonate
cGMP-compliant under the ISO 13485 standard
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.