Learn More
Gibco™ MEM, no glutamine, no phenol red

Description
Requires
Supplementation
MEM (Minimum Essential Medium) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This MEM is modified as follows:
Without: L-glutamine, HEPES, Phenol Red
The complete formulation is available.
Using MEM
MEM was developed by Harry Eagle, and was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow's MEM, MEM α, DMEM, and Temin's Modification. MEM is available with Earle's salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. This product is made with Earle's salts. MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (2.2 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Informations sur la commande
Shipping Condition: Room Temperature
Spécification
Spécification
| Cell Line | HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts |
| Cell Type | Primary Rat Astrocytes |
| Classification | Animal Origin-free |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | MEM (Minimum Essential Medium) |
| Serum Level | Standard Serum Supplementation |
| Sterility | Sterile-filtered |
| With Additives | Low Glucose |
| Without Additives | No Glutamine, No HEPES, No Phenol Red, No Sodium Pyruvate |
| Afficher plus |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.