missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM α, nucleosides, no phenol red
GREENER_CHOICE

Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negative cells.

£59.75 - £352.00

Specifications

Content And Storage Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Cell Line HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Cell Type Primary Rat Astrocytes
Classification Animal Origin-free
Concentration 1 X
View More Specs

Requires: Supplementation


Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11534476
GREENER_CHOICE
View Documents Promotion Details
Gibco™
41061029
500 mL
£59.75
500mL
Please to purchase this item. Need a web account? Register with us today!
15673854
GREENER_CHOICE
View Documents Promotion Details
Gibco™
41061037
10 x 500 mL
£352.00
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Minimum Essential Medium (MEM) α is widely used for mammalian cell culture as well as selection for transfected DHFR negative cells. MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells. We offer a variety of Gibco™ MEM α modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This MEM α is modified as follows:
With: Ribonucleosides, Deoxyribonucleosides, L-glutamine
Without: Phenol Red

The complete formulation is available.

Gibco™ MEM α is a modification of Minimum Essential Medium (MEM) that contains non-essential amino acids, sodium pyruvate, lipoic acid, vitamin B12, biotin, and ascorbic acid. MEM α is available without nucleosides for use as a selection medium for DG44 and other DHFR negative cells. This product is made with Earle’s salts.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

MEM α contains no proteins, lipids, or growth factors. Therefore, MEM α requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM α uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

Compliance

Gibco MEM α is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications

Specifications

Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Primary Rat Astrocytes
1 X
MEM α (Minimum Essential Medium α)
Sterile-filtered
No HEPES, No Phenol Red
cGMP-compliant under the ISO 13485 standard
Room Temperature
HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Animal Origin-free
Liquid
Standard Serum Supplementation
Low Glucose, Glutamine, Sodium Pyruvate, Ribonucleosides, Deoxyribonucleosides
Sustainable packaging
Gibco™
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 Months From Date of Manufacture

SDS
Documents

Documents

Product Certifications

For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.