Learn More
Gibco™ Gentamicin (50 mg/mL)
Water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea
£71.75 - £572.00
Specifications
Type | Gentamicin |
---|---|
Physical Form | Liquid |
Sterile Filtered | Yes |
Concentration | 50 mg/mL |
Validated Application | Prevention of Cell Culture Contamination |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12664735
|
Gibco™
15750060 |
10 mL |
£71.75
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15400604
|
Gibco™
15750078 |
10 x 10 mL | N/A | ||||||
12519079
|
Gibco™
15750078 |
10 x 10 mL |
£572.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Gentamicin sulfate is a water-soluble antibiotic originally purified from the fungus Micromonospora purpurea
- It acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition of protein synthesis and death in susceptible bacteria
- Gibco™ Gentamicin is effective against a wide variety of gram-positive and gram-negative bacteria, and is used to prevent the contamination of cell cultures by bacteria
- The recommended working concentration ranges from 0.5 to 50 μg /mL
- Store at 15°C to 30°C
Order Info
Shipping Condition: Room Temperature
Compliance
Manufactured at a cGMP compliant facility, which is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Gentamicin | |
Yes | |
Prevention of Cell Culture Contamination |
Liquid | |
50 mg/mL | |
Room Temperature |
Safety and Handling
Recommended Storage : Storage conditions: 15°C to 30°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture
- GENTAMICIN REAGENT SOLUTION (50 MG/ML), LIQUID
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : gentamicin sulphate
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.