Learn More
Gibco™ Neomycin Sulfate
Produced by Streptomyces fradiae
Brand: Gibco™ 21810031
Code : 50
Additional Details : Weight : 0.10000kg
Description
Polymyxin B Sulfate acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition of protein synthesis and death in susceptible bacteria. Gibco Neomycin Sulfate is used for the prevention of bacterial contamination of cell cultures. This antibiotic is highly active against gram-negative bacteria.
Cell Culture, Cloning, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Sf9 & Sf21 Cell Culture, Transformation
Order Info
Shipping Condition: Room Temperature
Specifications
Storage conditions: 15 to 30°C Shipping conditions: Ambient Shelf life: 24 months from date of manufacture |
|
Powder | |
Non-sterile | |
100 g |
Mammalian Cell Culture, Insect Cell Culture | |
Antibiotic | |
Bacterial Selection | |
Room Temperature |
Safety and Handling
- Neomycin Sulphate
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Neomycin sulphate
For Research Use Only. Not for use in diagnostic procedures.