missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Polymyxin B Sulfate
Polymyxin B Sulfate is produced by Bacillus polymyxa
Brand: Gibco™ 21850029
123.17 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
This item is not returnable.
View return policy
Description
Polymyxin B Sulfate works by binding to the cellular membrane and interfering with its permeability. Gibco™ Polymyxin B Sulfate is used for the prevention of bacterial contamination of cell cultures. This antibiotic is highly active against gram-negative bacteria.
- Available as 25 MU powder
- Recommended working concentration is 100 units/mL
Cloning, Transformation
Order Info
Shipping Condition: Room Temperature
Specifications
Storage conditions: 15 to 30°C Shipping conditions: Ambient Shelf life: 24 months from date of manufacture |
|
Powder | |
Non-sterile | |
25 MU |
100 Units/mL | |
Antibiotic | |
Bacterial Selection | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.