Learn More
Gibco™ Sf-900™ II SFM, no methionine, no cystine
Optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells
Brand: Gibco™ 21012026
Code : 50
Additional Details : Weight : 0.00100kg
Description
Sf-900 II SFM (without methionine and cystine) is a serum-free, protein-free insect cell culture medium optimized for large-scale production of recombinant proteins expressed using the baculovirus expression vector system (BEVS). This medium supports growth of other lepidopteran cell lines.
- Superior long-term, high-density growth
- Optimized for recombinant protein production
- Serum-free, protein-free, ready-to-use formulation
- Scalable in bioreactors
Bioproduction, Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture, Viral Vaccine Production
Order Info
Shipping Condition: Room temperature
Specifications
Storage conditions: 2°C to 8°C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Insect Cell | |
Liquid | |
Serum-free | |
Gibco™, Sf-900™ | |
S. frugiperda, Spodoptera frugiperda |
Sf21, Sf9 | |
Protein-free, Serum-free | |
Insect Cell Serum Free Medium (SFM) | |
Sustainable packaging | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.