Learn More
Gibco™ Soybean Trypsin Inhibitor, powder
Trypsin inhibited at a molar ratio of 1:1 and chymotrypsin and plasmin to lesser extent
Brand: Gibco™ 17075029
89.78 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
Also inhibits other serine proteases. Has an activity greater than 7,000 BAEE units per mg. A 0.5mg/mL Soybean Trypsin Inhibitor stock solution should be made by dissolving nonsterile powder in 0.01M Borate Buffer or Dulbecco's Phosphate Buffered Saline (DPBS), without calcium and magnesium. Filter sterilize the stock solution and store for up to a week at 2 - 8°C.
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: -5 to -30°C Shipping conditions: Frozen |
|
Frozen, powder | |
Non-sterile | |
1 g |
Mammalian Cells | |
Trypsin inhibitor | |
No Calcium, No Magnesium | |
Dry Ice |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.