missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Newborn Calf Serum, heat inactivated, New Zealand origin
Secure sera for cell culture with least viral risk sera from BSE-free regions.
Brand: Gibco™ 26010066
This item is not returnable.
View return policy
Description
Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.- Sera Category: Secure
- Origin: New Zealand
- Collected from calves 14 days old or less
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Cattle/Bovine | |
Liquid | |
Newborn Calf Bovine Serum | |
Heat-inactivated | |
Frozen |
Newborn (14 days or less) | |
New Zealand | |
Bottle | |
100 mL | |
Gibco™ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction