missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Newborn Calf Serum, New Zealand origin
General stem cell culture media and reagents; sera for specialty research and specific assays
£27.81 - £190.00
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
---|---|
Age | Newborn (20 days or less) |
Species | Cattle/Bovine |
Country of Origin | New Zealand |
Format | Bottle |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11590506
|
Gibco™
16010167 |
100 mL |
£27.81
100mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11580506
|
Gibco™
16010159 |
500 mL |
£100.00
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17862964
|
Gibco™
16010142 |
1000 mL |
£190.00
1 L |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sera Category: Secure
- Origin: New Zealand
- Collected from calves 14 days old or less
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Cattle/Bovine | |
Bottle | |
Sterile | |
Frozen |
Newborn (20 days or less) | |
New Zealand | |
Standard (Sterile-filtered) | |
Gibco™ |