missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PB-MAX™ Karyotyping Medium
GREENER_CHOICE

Product Code. 10722057
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10722057

Brand: Gibco™ 12557013

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Fully supplemented medium developed for short-term culture of peripheral blood lymphocytes for cytogenetic studies and in vitro diagnostic procedures

  • Ready-to-use format
  • Quality and performance testing
  • Classic formulation
  • Sustainable packaging

Ready-to-use format

1X medium is a frozen, ready to use upon thawing. No supplementation is required. Thawed medium can be stored at 2° to 8°C for up to 14 days.

Quality and performance testing

Every lot of is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance. Peripheral blood lymphocytes from a normal adult donor are cultured for 72 hours in Gibco PB-MAX Karyotyping Medium before measuring the mitotic index and chromosome banding resolution. In addition, each lot is tested for sterility, pH, osmolality, and mycoplasma.

Classic formulation

Gibco PB-MAX Karyotyping Medium is an optimized RPMI 1640 medium, supplemented with Fetal Bovine Serum (FBS), L-glutamine, and phytohemagglutinin (PHA). Gibco PB-MAX Karyotyping Medium is based on formulations referenced in the Association for Cytogenetic Technologists Laboratory Manual for the culture of peripheral blood lymphocytes for cytogenetic analysis.

Cell Analysis, Cell Culture, Cell Proliferation, Cell Viability, Proliferation and Function, Clinical, Clinical and Translational Research, Cytogenetics, Karyotyping, Mammalian Cell Culture

Order Info

Shipping Condition: Dry Ice

Compliance

Manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Karyotyping Medium
Color Red
Concentration 1 X
For Use With (Application) Cell Viability; Proliferation & Function; Clinical & Translational Research; Mammalian Cell Culture
Physical Form Liquid
Product Line PB-MAX
Quantity 100 mL
Sterility Sterile-filtered
Includes L-Glutamine

For in vitro diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.