missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PBS (10X), pH 7.4
GREENER_CHOICE

Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

£27.07

Specifications

Additive No Sodium Pyruvate
Classification Animal Origin-Free
Color White
Content And Storage Storage conditions: 15°C to 30°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture
Dilution 10 X
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12579099
GREENER_CHOICE
View Documents
Gibco™
70011044
500 mL
£27.07
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution (see protocol for details).

We offer a variety of Gibco™ PBS formulations for a range of cell culture applications.

This PBS is manufactured as follows:

Without: Calcium, Magnesium, Phenol Red

The complete formulation is available.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

No Sodium Pyruvate
White
10 X
Phosphate Buffered Saline (PBS)
Mammalian Cell Culture
7.4
Room Temperature
Liquid
Animal Origin-Free
Storage conditions: 15°C to 30°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture
Gibco™
Phosphate Buffered Saline
2800 - 3100 mOsm/kg
No Phenol Red
Sterile-filtered
cGMP-compliant under the ISO 13485 standard
Videos
SDS
Documents

Documents

Product Certifications

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.