missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PBS, pH 7.2
GREENER_CHOICE

Product Code. p-4924958
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 12549079

Brand: Gibco™ 20012027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gibco PBS, pH 7.2, is a balanced salt solution widely used in various cell culture applications to maintain a stable pH and osmotic balance, making it ideal as a wash, transport, and dilution buffer.

Gibco Phosphate-Buffered Saline (PBS), pH 7.2, is a balanced salt solution widely used in various cell culture applications. It is designed to maintain a stable pH and osmotic balance, making it ideal for washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

Key features

  • Without calcium and magnesium—the absence of calcium and magnesium is crucial for applications requiring the rinsing of chelators from the culture before cell dissociation
  • Without phenol red—the lack of phenol red ensures that there is no interference with optical measurements and prevents potential effects on cell physiology

Applications

  • Cell washing—ideal for washing cells before dissociation to remove any residual enzymes or chelators
  • Sample transport—suitable for transporting cells or tissue samples while maintaining their viability
  • Cell counting—can be used to dilute cells for accurate counting
  • Reagent preparation—provides a stable environment for preparing various reagents used in cell culture

Why choose PBS vs. DPBS

  • Simplicity—PBS is a simpler formulation compared to Dulbecco's Phosphate-Buffered Saline (DPBS), which includes additional components like potassium chloride and may be formulated with or without calcium, magnesium, glucose, and pyruvate. This simplicity makes PBS a versatile choice for general cell culture applications.
  • Specific applications—PBS is preferred for applications where the presence of potassium or other additives in DPBS might interfere with experimental outcomes

Why choose pH 7.2 vs. pH 7.4 PBS

  • Experimental requirements—the choice between pH 7.2 and pH 7.4 depends on the specific requirements of your cell culture system. pH 7.2 is often used for certain cell types and experimental conditions where a slightly more acidic environment is beneficial.
  • Buffering capacity—pH 7.2 provides a slightly different buffering capacity, which can be crucial for maintaining the desired physiological conditions in specific experimental setups

Why choose Gibco PBS, pH 7.2

  • Quality and consistency—Gibco products are manufactured in cGMP, ISO-certified facilities, ensuring the highest quality and consistency. Each batch undergoes rigorous testing for osmolality, pH levels, sterility, and the presence of endotoxins.
  • Track record—Gibco cell culture products have a long-standing reputation for excellence in the field of cell culture, with numerous publications citing the use of Gibco products. This track record of reliability and performance makes Gibco PBS the preferred choice for researchers worldwide.

Gibco PBS, pH 7.2 is a reliable and consistent solution for a wide range of applications. Choose Gibco products for your cell culture needs and benefit from our decades of expertise and innovation in the field.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Color White
Concentration 1 X
For Use With (Application) Mammalian Cell Culture
pH 7.2
Physical Form Liquid
Product Line Gibco
Quantity 500 mL
Solution Type Phosphate Buffered Saline
Sterility Sterile-filtered
Includes No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Osmolality 280 - 320 mOsm/kg
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.