missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse PDGF-BB Recombinant Protein
Recombinant Mouse Platelet Derived Growth Factor-BB (PDGF-BB) is a bioactive protein intended for use in cell culture applications
£161.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Gene ID (Entrez) | 18591 |
Gene Alias | PDGF-BB |
Species | Mouse |
Content And Storage | Store in refrigerator (2–8°C). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10049222
|
Gibco™
PMG0044 |
10μg | |||||||
Description
Members of the PDGF family are mitogenic factors for cells of mesenchymal origin.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose-dependent proliferation of BALB/c 3T3 cells
Specifications
Cell Culture | |
PDGF-BB | |
Store in refrigerator (2–8°C). | |
E. coli | |
Recombinant, Ligand | |
PDGFs (Platelet-Derived Growth Factors) | |
Gibco™ |
18591 | |
Mouse | |
Wet Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Immunology, Neurobiology, Stem Cell Research, Angiogenesis, Oncology |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.