missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Penicillin-Streptomycin (10,000 U/mL)

Product Code. p-4918413
Click to view available options
Quantity:
20 mL
100 mL
20 x 100 mL
Unit Size:
100mL
20mL
Pack of 20
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11568876

Brand: Gibco™ 15140148

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

For prevention of bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria

Penicillin was originally purified from fungi Penicillium that acts by interfering directly with turnover of bacterial cell wall and indirectly by triggering release of enzymes that alter the cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to 30S subunit of bacterial ribosome, leading to inhibition of protein synthesis and death in susceptible bacteria.

  • Contains 10,000 units/mL of penicillin
  • 10,000μg/mL of streptomycin in a 10 mM citrate buffer (for pH stability)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: -5 to -20°C
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture
Concentration 100 X
Form Liquid
Product Type Antibiotic
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
For Use With (Application) Prevention of Cell Culture Contamination
Quantity 20 mL
Shipping Condition Dry Ice
compliance-icons
Product Identifier
  • Penicillin-Streptomycin, liquid
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : streptomycin sulphate

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.