missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Penicillin-Streptomycin-Glutamine (100X)
GREENER_CHOICE

For prevention from bacterial contamination in cell cultures due to their effective combined action against gram-positive and gram-negative bacteria.

Brand:  Gibco™ 10378016

22.64 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.



Product Code. 12090216

  • £29.26 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Penicillin was originally purified from the fungi Penicillium and acts by interfering directly with the turnover of the bacteria cell wall and indirectly by triggering the release of enzymes that alter cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to 30S subunit of the bacterial ribosome, leading to the inhibition of protein synthesis and death in susceptible bacteria.

  • Available as 100 mL liquid
  • Solution contains 10,000 units of penicillin, 10,000μg of streptomycin, and 29.2 mg/mL of L-glutamine in a 10 mM citrate buffer (for pH stability)

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: -5°C to -20°C
Shipping conditions: Dry ice
Shelf life: 12 months from date of manufacture
Liquid
Sterile-filtered
Prevention of Cell Culture Contamination
Dry Ice
100 X, 50 mg/mL
Antibiotic-L-Glutamine
Glutamine
100 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Penicillin-Streptomycin-Glutamine (100X)

Signal Word
  • Warning

Hazard Category
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361-Suspected of damaging fertility or the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Streptomycin sulfate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.