missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Phytohemagglutinin, M form (PHA-M)

This extract is capable of inducing blastogenesis in vitro in various mammalian mononuclear cells when used at a concentration of 1 to 2% v/v

Brand:  Gibco™ 10576015

Product Code. 11570356

  • £35.87 / 10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

  • Made from a crude extract of the red kidney bean Phaseolus vulgaris
  • Each lot is performance tested at 1.5% using peripheral blood lymphocytes from a normal adult donor cultured for 72 hours in Gibco™ RPMI 1640 Medium supplemented with Fetal Bovine Serum (FBS), heparin, antibiotics, and L-glutamine
  • Manufactured at a cGMP compliant facility, located in Grand Island, New York, registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards

Baculovirus Production and Titering, Cell Culture, Insect Cell Culture, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: -5°C to -20°C. Protect from light.
Shipping conditions: Dry ice
Shelf life: 24 months from date of manufacture
Liquid
Sterile-filtered
Dry Ice
Mammalian Cells
Phytohemagglutinin
10 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2562624 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number10576015
Lot Number2876834 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number10576015
Lot Number2905318 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number10576015
Lot Number2337679 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number10576015
Lot Number2876838 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number10576015
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Phytohemagglutinin, M form (PHA-M) > 1 x 10mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.