missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
---|---|
Age | 6 months or less |
Species | Rabbit |
Country of Origin | United States |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11550516
|
Gibco™
16120099 |
100 mL |
£37.70
100mL |
In Stock
Log in to see stock. |
|||||
11560516
|
Gibco™
16120107 |
500 mL |
£150.00
500mL |
In Stock
Log in to see stock. |
|||||
Description
- Sera Category: Specialty
- Origin: United States
- Age: 6 months or less
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Rabbit | |
Liquid | |
Standard (Sterile-filtered) | |
Gibco™ |
6 months or less | |
United States | |
Rabbit Serum | |
Sterile | |
Frozen |
For in vitro diagnostic
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title