missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ RPMI, no methionine
GREENER_CHOICE

Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer.

Brand:  Gibco™ A1451701

Product Code. 12857552

  • £89.25 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 medium has since been found suitable for a variety of mammalian cells including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of Gibco™ RPMI 1640 modifications for a range of cell culture applications. Find the right formulation using our media selector tool.

This RPMI medium is modified as follows:
With: L-glutamine, Phenol Red
Without: Methionine, HEPES

Gibco™ RPMI 1640 medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations.

Product Intended Use
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

RPMI 1640 medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 medium requires supplementation, commonly with 10% fetal bovine serum (FBS). RPMI 1640 medium uses a sodium bicarbonate buffer system (2.0 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Certifications

ISO 13485

TRUSTED_SUSTAINABILITY
Specifications

Specifications

No Methionine
HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas
Animal Origin-free
Liquid
Standard Serum Supplementation
Glutamine, Phenol Red, Sodium Bicarbonate
Mammalian Cell Culture
cGMP-compliant under the ISO 13485 standard
500 mL
Room Temperature
Storage conditions: 2-8 Deg;C. Protect from light



Shipping conditions: Ambient



Shelf life: 12 months from date of manufacture
Leukemic Cells
1 X
RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
Sterile-filtered
No HEPES, No Sodium Pyruvate
Sustainable packaging
Gibco™
RUO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions